Assay Details
Target Gene Details
Entrez Gene ID: | 76507 |
Gene Name: | amine oxidase, copper-containing 1 |
Gene Aliases: |
1600012D06Rik, Abp1, DAO |
Location: |
Chr.6:48895255-48909188 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aoc1 | NM_001161621.1 | 4 | 2313 | NP_001155093.1 |
NM_001161622.1 | 5 | 2216 | NP_001155094.1 | |
NM_029638.2 | 5 | 2155 | NP_083914.2 | |
AK005423.1 | 3 | 453 | BAB24017.1 | |
AK144567.1 | 5 | 2155 | ||
AK146078.1 | 4 | 2314 | BAE26882.1 | |
BC021880.1 | 5 | 2156 | AAH21880.1 | |
BC022627.1 | 4 | 1180 | AAH22627.1 | |
BC034215.1 | 5 | 2127 | AAH34215.1 |
More Information
Set Membership: |
Intragenic Exonic |