Assay Details
Target Gene Details
Entrez Gene ID: | 14910 |
Gene Name: | gene trap ROSA 26, Philippe Soriano |
Gene Aliases: |
AV258896, Gtrgeo26, Gtrosa26, R26, ROSA26 |
Location: |
Chr.6:113067428-113077244 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gt(ROSA)26Sor | NR_027010.1 | 3 | 261 | |
AK021028.1 | 3 | 208 | ||
BY727583.1 | 3 | 206 | ||
U83175.1 | 3 | 300 |
Target Gene Details
Entrez Gene ID: | 14911 |
Gene Name: | THUMP domain containing 3 |
Gene Aliases: |
AW556087, Gt(ROSA)26asSor, Gtrosa26as |
Location: |
Chr.6:113046252-113068278 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Thumpd3 | NM_008188.2 | 10 | 1559 | NP_032214.1 |
AK142140.1 | 10 | 1578 | BAE24945.1 | |
BC002024.1 | 10 | 1561 | AAH02024.1 | |
BC012688.1 | 9 | 1507 | AAH12688.1 | |
BC046800.1 | 10 | 1559 | AAH46800.1 | |
U83176.1 | 10 | 1553 | AAC60384.1 |
More Information
Set Membership: |
Intragenic Exonic |