Assay Details
Target Gene Details
Entrez Gene ID: | 114142 |
Gene Name: | forkhead box P2 |
Gene Aliases: |
2810043D05Rik, AI449000, CAG-16, D0Kist7 |
Location: |
Chr.6:14901226-15441977 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Foxp2 | NM_053242.4 | 1 | 341 | NP_444472.2 |
AK051668.1 | 1 | 303 | BAC34711.1 | |
AK080549.1 | 1 | 212 | ||
AK082361.1 | BAC38477.1 | |||
AK085080.1 | 1 | 240 | ||
CN526766.1 | 1 | 342 |