Assay Details
Target Gene Details
Entrez Gene ID: | 22377 |
Gene Name: | WW domain binding protein 1 |
Gene Aliases: |
AI255152 |
Location: |
Chr.6:83119044-83121663 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Wbp1 | NM_001083922.1 | 1 | 218 | NP_001077391.1 |
NM_001083923.1 | 1 | 218 | NP_001077392.1 | |
NM_016757.2 | 1 | 218 | NP_058037.2 | |
AK076948.1 | 1 | 153 | BAC36533.1 | |
AK152822.1 | 1 | 220 | ||
AK170907.1 | 1 | 188 | ||
AK179139.1 | 1 | 71 | ||
AK184383.1 | 1 | 84 | ||
BC005666.1 | 1 | 334 | AAH05666.1 | |
BC019889.1 | 1 | 142 | AAH19889.1 | |
BE860581.1 | 1 | 52 | ||
BY159385.1 | 1 | 218 | ||
CX205200.1 | 1 | 189 | ||
U40825.1 | 1 | 180 | AAB40892.1 |