Assay Details
Target Gene Details
Entrez Gene ID: | 27055 |
Gene Name: | FK506 binding protein 9 |
Gene Aliases: |
1810014L23, AW549641, FKBP-63, FKBP-9, FKBP60, FKBP63, FKBP65RS |
Location: |
Chr.6:56832059-56879360 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fkbp9 | NM_012056.2 | 10 | 2128 | NP_036186.2 |
AF090334.1 | 10 | 2054 | AAC72964.1 | |
AF279263.1 | 10 | 2106 | AAF79215.1 | |
AK007499.1 | 8 | 1518 | BAB25071.1 | |
AK188764.1 | 1 | 210 | ||
AK199007.1 | 1 | 210 | ||
AK204930.1 | 1 | 210 | ||
AK211458.1 | 1 | 210 | ||
AK218915.1 | 1 | 210 | ||
BC026133.1 | 10 | 2068 | AAH26133.1 | |
BC043129.1 | 10 | 2128 | AAH43129.1 |