Assay Details
Target Gene Details
Entrez Gene ID: | 26910 |
Gene Name: | folliculogenesis specific basic helix-loop-helix |
Gene Aliases: |
bHLHc8 |
Location: |
Chr.6:86010320-86020996 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Figla | NM_012013.1 | 3 | 477 | NP_036143.1 |
JN700518.1 | 3 | 437 | ||
U91840.1 | 3 | 477 | AAB97139.1 |
More Information
Set Membership: |
Intragenic Exonic |