Assay Details
Target Gene Details
Entrez Gene ID: | 75010 |
Gene Name: | transmembrane BAX inhibitor motif containing 7 |
Gene Aliases: |
4930403J02Rik, 4930500J03Rik, 4930511M11Rik, Lgf5, Tmbim1b |
Location: |
Chr.5:3657004-3680677 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tmbim7 | NM_029141.4 | 1 | 207 | NP_083417.1 |
NM_029154.1 | 1 | 207 | NP_083430.1 | |
AK015666.1 | 1 | 207 | BAB29920.1 | |
AK015761.1 | BAB29963.1 | |||
AK019559.1 | ||||
BC115681.2 | 1 | 65 |