Assay Details
Target Gene Details
Entrez Gene ID: | 70832 |
Gene Name: | RIKEN cDNA 4921504A21 gene |
Gene Aliases: |
- |
Location: |
Chr.5:19202368-19226555 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
4921504A21Rik | NR_102341.1 | 1 | 277 | |
NR_102342.1 | 1 | 277 | ||
AK046351.1 | 1 | 277 |
Target Gene Details
Entrez Gene ID: | 50791 |
Gene Name: | membrane associated guanylate kinase, WW and PDZ domain containing 2 |
Gene Aliases: |
AIP-1, Acvri1, Acvrinp1, Acvrip1, Magi-2, S-SCAM, mKIAA0705 |
Location: |
Chr.5:19223283-20704792 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Magi2 |