Assay Details
Target Gene Details
Entrez Gene ID: | 230908 |
Gene Name: | TAR DNA binding protein |
Gene Aliases: |
1190002A23Rik, C85084, TDP-43, Tdp43 |
Location: |
Chr.4:148612382-148626996 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tardbp | NM_001003898.3 | 7 | 2228 | NP_001003898.1 |
NM_001003899.2 | 7 | 2219 | NP_001003899.1 | |
NM_001008545.2 | 7 | 2246 | NP_001008545.1 | |
NM_001008546.2 | 7 | 2158 | NP_001008546.1 | |
NM_145556.4 | 6 | 3187 | NP_663531.1 | |
NR_027864.1 | 8 | 1507 | ||
AK032199.1 | 6 | 2957 | BAC27753.1 | |
AK045495.1 | 8 | 1277 | BAC32395.1 | |
AK083312.1 | 6 | 2961 | BAC38861.1 | |
AK133207.1 | 6 | 2964 | BAE21557.1 |