Assay Details
Target Gene Details
Entrez Gene ID: | 230161 |
Gene Name: | acyl-coenzyme A amino acid N-acyltransferase 1 |
Gene Aliases: |
AI132189 |
Location: |
Chr.4:49443532-49481312 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Acnat1 | NM_001164565.1 | 3 | 880 | NP_001158037.1 |
DQ469311.1 | 3 | 880 |
More Information
Set Membership: |
Intragenic Exonic |