Assay Details
Target Gene Details
Entrez Gene ID: | 74761 |
Gene Name: | matrix-remodelling associated 8 |
Gene Aliases: |
1200013A08Rik, 1700095D18Rik, AI131686, Asp3, Dicam |
Location: |
Chr.4:155839680-155844102 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mxra8 | NM_024263.4 | 5 | 1017 | NP_077225.4 |
AB040488.1 | 5 | 937 | BAB68501.1 | |
AB052097.1 | 5 | 895 | BAD05133.1 | |
AK004732.1 | 5 | 900 | BAB23514.1 | |
AK182996.1 | 1 | 103 | ||
BC026438.1 | 5 | 939 | AAH26438.1 | |
EU849596.1 | 5 | 855 | ||
EU849597.1 | 5 | 846 |