Assay Details
Target Gene Details
Entrez Gene ID: | 56455 |
Gene Name: | dynein light chain LC8-type 1 |
Gene Aliases: |
Dlc8, Dnclc1, Pin |
Location: |
Chr.5:115297110-115300990 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dynll1 | NM_019682.4 | 3 | 875 | NP_062656.3 |
AK082923.1 | 3 | 820 | BAC38691.1 | |
AK147977.1 | 3 | 820 | BAE28262.1 |
More Information
Set Membership: |
Intragenic Exonic |