Assay Details
Target Gene Details
Entrez Gene ID: | 56221 |
Gene Name: | chemokine (C-C motif) ligand 24 |
Gene Aliases: |
CKb-6, MPIF-2, Scya24 |
Location: |
Chr.5:135569937-135573122 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccl24 | NM_019577.4 | 3 | 773 | NP_062523.1 |
AK157068.1 | 3 | 774 | BAE33951.1 |
More Information
Set Membership: |
Intragenic Exonic |