Assay Details
Target Gene Details
Entrez Gene ID: | 18786 |
Gene Name: | phospholipase A2, activating protein |
Gene Aliases: |
2410007N06, AI536418, AU018445, AW208417, D4Ertd618e, PLA2P, PLAP, Ufd3 |
Location: |
Chr.4:94565139-94603247 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Plaa | NM_172695.2 | 14 | 2836 | NP_766283.2 |
AK085027.1 | 4 | 2283 | ||
BC139355.1 | 14 | 2835 | ||
BC139356.1 | 14 | 2835 |
More Information
Set Membership: |
Intragenic Exonic |