Assay Details
Target Gene Details
Entrez Gene ID: | 73471 |
Gene Name: | RIKEN cDNA 1700066N21 gene |
Gene Aliases: |
- |
Location: |
Chr.5:87908588-87979351 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
1700066N21Rik | NR_045924.1 | |||
AK006908.1 |
Target Gene Details
Entrez Gene ID: | 20599 |
Gene Name: | submaxillary gland androgen regulated protein 3A |
Gene Aliases: |
AA589601, MSG1, MSG3, Smr1, Smr3 |
Location: |
Chr.5:87969504-88008534 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smr3a |