Assay Details
Target Gene Details
Entrez Gene ID: | 22178 |
Gene Name: | tyrosinase-related protein 1 |
Gene Aliases: |
Oca3, TRP-1, TRP1, Tyrp, b, brown, isa |
Location: |
Chr.4:80834208-80851736 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tyrp1 | NM_001282014.1 | 3 | 761 | NP_001268943.1 |
NM_001282015.1 | 3 | 755 | NP_001268944.1 | |
NM_031202.3 | 3 | 776 | NP_112479.1 | |
AK148332.1 | 3 | 761 | BAE28487.1 | |
AK148370.1 | 3 | 667 | BAE28511.1 | |
AK148441.1 | 3 | 667 | BAE28554.1 | |
AK184972.1 | 2 | 259 | ||
AK190422.1 | 2 | 259 | ||
AK191393.1 | 2 | 129 | ||
AK191462.1 | 2 | 259 | ||
AK191697.1 | 2 | 258 | ||
AK192907.1 | 2 | 257 | ||
AK192940.1 | 2 | 259 | ||
AK195260.1 | 2 | 224 | ||
AK199454.1 | 2 | 216 | ||
AK200238.1 | 2 | 259 | ||
AK201835.1 | 2 | 260 | ||
AK202735.1 | 2 | 259 | ||
AK202956.1 | 1 | 77 | ||
AK204438.1 | 2 | 202 | ||
AK204752.1 | 2 | 259 | ||
AK207360.1 | 2 | 226 | ||
AK214203.1 | ||||
BC076598.1 | 3 | 759 | AAH76598.1 | |
X03687.1 | 3 | 661 | CAA27323.1 |
More Information
Set Membership: |
Intragenic Exonic |