Assay Details
Target Gene Details
Entrez Gene ID: | 231842 |
Gene Name: | archaelysin family metallopeptidase 1 |
Gene Aliases: |
5330426I05, 6530401C20Rik |
Location: |
Chr.5:140724127-140753312 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Amz1 | NM_173405.2 | 8 | 2003 | NP_775581.1 |
AK078332.1 | 8 | 2003 | BAC37222.1 | |
AK170176.1 | 7 | 1977 | BAE41617.1 | |
BC046428.1 | 3 | 971 | AAH46428.1 | |
BC129841.1 | 6 | 1786 | ||
BC129842.1 | 6 | 1738 |