Assay Details
Target Gene Details
Entrez Gene ID: | 14620 |
Gene Name: | gap junction protein, beta 3 |
Gene Aliases: |
Cnx31, Cx31, D4Wsu144e, Gjb-3 |
Location: |
Chr.4:127325235-127330836 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gjb3 | NM_001160012.1 | 2 | 1358 | NP_001153484.1 |
NM_008126.2 | 2 | 1073 | NP_032152.1 | |
AK083716.1 | 2 | 1115 | BAC39002.1 | |
BC024387.1 | 2 | 1051 | AAH24387.1 | |
X63099.1 | 2 | 1109 | CAA44813.1 |
More Information
Set Membership: |
Intragenic Exonic |