Assay Details
Target Gene Details
Entrez Gene ID: | 14620 |
Gene Name: | gap junction protein, beta 3 |
Gene Aliases: |
Cnx31, Cx31, D4Wsu144e, Gjb-3 |
Location: |
Chr.4:127325235-127330836 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gjb3 | NM_001160012.1 | 1 | 286 | NP_001153484.1 |
AK083716.1 | 1 | 43 | BAC39002.1 | |
AY390400.1 | 1 | 294 | AAQ93033.1 | |
BC024387.1 | AAH24387.1 | |||
BY027476.1 | 1 | 288 | ||
CJ084102.1 | 1 | 291 |
More Information
Set Membership: |
Intragenic Exonic |