Assay Details
Target Gene Details
Entrez Gene ID: | 231842 |
Gene Name: | archaelysin family metallopeptidase 1 |
Gene Aliases: |
5330426I05, 6530401C20Rik |
Location: |
Chr.5:140724127-140753312 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Amz1 | NM_173405.2 | 3 | 583 | NP_775581.1 |
AJ879912.1 | 1 | 224 | CAI53757.1 | |
AK078332.1 | 3 | 583 | BAC37222.1 | |
AK170176.1 | 2 | 557 | BAE41617.1 | |
BC129841.1 | 1 | 366 | ||
BC129842.1 | 1 | 366 |