Assay Details
Target Gene Details
Entrez Gene ID: | 231642 |
Gene Name: | alkB, alkylation repair homolog 2 (E. coli) |
Gene Aliases: |
9530023G02, AU016977, Abh2, mABH2 |
Location: |
Chr.5:114123926-114130546 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Alkbh2 | NM_175016.2 | 4 | 796 | NP_778181.2 |
AK079195.1 | 4 | 633 | BAC37576.1 | |
BC062188.1 | 4 | 796 | AAH62188.1 |
More Information
Set Membership: |
Intragenic Exonic |