Assay Details
Target Gene Details
Entrez Gene ID: | 230145 |
Gene Name: | UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 12 |
Gene Aliases: |
9130206E10, A630062B03Rik |
Location: |
Chr.4:47091893-47123042 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Galnt12 | NM_172693.3 | 10 | 2122 | NP_766281.1 |
AK033638.1 | 10 | 2123 | BAC28401.1 |
More Information
Set Membership: |
Intragenic Exonic |