Assay Details
Target Gene Details
Entrez Gene ID: | 14086 |
Gene Name: | fascin homolog 1, actin bundling protein (Strongylocentrotus purpuratus) |
Gene Aliases: |
AI663989, Fan1, fascin-1 |
Location: |
Chr.5:142960355-142973189 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fscn1 | NM_007984.2 | 1 | 616 | NP_032010.2 |
AK083935.1 | 1 | 616 | ||
AK167670.1 | 1 | 616 | BAE39719.1 | |
BC010338.1 | 1 | 602 | ||
BC037137.1 | 1 | 613 | ||
BC052408.1 | 1 | 605 | AAH52408.1 | |
L33726.1 | 1 | 600 | AAB41126.1 |
More Information
Set Membership: |
Intragenic Exonic |