Assay Details
Target Gene Details
Entrez Gene ID: | 69663 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 51 |
Gene Aliases: |
2310061O04Rik, AA537002, C81583, R75505 |
Location: |
Chr.5:110653434-110660496 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx51 | NM_027156.3 | 15 | 3387 | NP_081432.2 |
BC060646.1 | 15 | 3364 | AAH60646.1 |