Assay Details
Target Gene Details
Entrez Gene ID: | 56079 |
Gene Name: | astrotactin 2 |
Gene Aliases: |
1d8, Astnl, bM452J22.1 |
Location: |
Chr.4:65380803-66404530 on Build GRCm38 |
Assay Gene Location: | Within Intron 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Astn2 | NM_019514.3 | NP_062387.2 | ||
NM_207109.2 | NP_996992.1 | |||
AF116573.1 | AAF14356.1 | |||
AK041160.1 | BAC30843.1 |
Target Gene Details
Entrez Gene ID: | 69807 |
Gene Name: | tripartite motif-containing 32 |
Gene Aliases: |
1810045E12Rik, 3f3, BBS11, Zfp117 |
Location: |
Chr.4:65604986-65616240 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trim32 | NM_001161782.1 | 2 | 603 | NP_001155254.1 |
NM_053084.2 | 2 | 606 | NP_444314.2 | |
AF347694.1 | 2 | 574 | AAO13297.1 | |
AK154028.1 | 2 | 571 | BAE32325.1 | |
AK166358.1 | 2 | 558 | BAE38729.1 | |
AX775777.1 | 2 | 586 | CAE11588.1 | |
BC034104.1 | 2 | 545 | AAH34104.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |