Assay Details
Target Gene Details
Entrez Gene ID: | 67096 |
Gene Name: | methylmalonic aciduria cblC type, with homocystinuria |
Gene Aliases: |
1810037K07Rik |
Location: |
Chr.4:116702434-116708385 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mmachc | NM_025962.3 | 4 | 740 | NP_080238.2 |
AK007725.1 | 4 | 675 | BAB25214.1 | |
AK012761.1 | 4 | 677 | BAB28451.1 | |
AK084194.1 | 4 | 691 | BAC39135.1 | |
BC054756.1 | 4 | 679 | AAH54756.1 |