Assay Details
Target Gene Details
Entrez Gene ID: | 106021 |
Gene Name: | topoisomerase I binding, arginine/serine-rich |
Gene Aliases: |
AW105885, LUN, TP53BPL, p53BP3/LUN |
Location: |
Chr.4:40259606-40269841 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Topors | NM_134097.3 | 3 | 949 | NP_598858.2 |
AB072395.1 | 3 | 906 | BAB69457.1 | |
AB104865.1 | 3 | 934 | BAC65157.1 | |
AK044564.1 | 3 | 927 | BAC31981.2 | |
AK134075.1 | 3 | 945 | BAE22003.1 | |
AK140250.1 | 3 | 950 | BAE24298.1 | |
AK143025.1 | 3 | 930 | BAE25253.1 | |
AK153743.1 | 3 | 936 | BAE32164.1 | |
BC019421.1 | 3 | 907 | ||
BC037141.1 | 3 | 927 | AAH37141.1 | |
BC040797.1 | 3 | 937 | AAH40797.1 |
More Information
Set Membership: |
Intragenic Exonic |