Assay Details
Target Gene Details
Entrez Gene ID: | 74087 |
Gene Name: | solute carrier family 7, (cationic amino acid transporter, y+ system) member 13 |
Gene Aliases: |
0610009O04Rik, AGT-1, AGT1, XAT2 |
Location: |
Chr.4:19818727-19842213 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc7a13 | NM_028746.3 | 3 | 954 | NP_083022.1 |
AB072352.1 | 3 | 937 | BAC00494.1 | |
AJ417662.1 | 3 | 903 | CAD10394.1 | |
AK128901.1 | 3 | 954 | ||
AK143768.1 | 3 | 956 | BAE25532.1 | |
AK165403.1 | 3 | 955 | ||
BC014684.1 | 3 | 927 | AAH14684.1 |
More Information
Set Membership: |
Intragenic Exonic |