Assay Details
Target Gene Details
Entrez Gene ID: | 20310 |
Gene Name: | chemokine (C-X-C motif) ligand 2 |
Gene Aliases: |
CINC-2a, GROb, Gro2, MIP-2, MIP-2a, Mgsa-b, Mip2, Scyb, Scyb2 |
Location: |
Chr.5:90903899-90905938 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cxcl2 | NM_009140.2 | 4 | 450 | NP_033166.1 |
AK137628.1 | 4 | 478 | BAE23436.1 | |
AK150450.1 | 4 | 358 | BAE29571.1 | |
AK155458.1 | 1 | 1434 | ||
AK155874.1 | 3 | 1227 | BAE33476.1 | |
AK155916.1 | 4 | 477 | BAE33502.1 | |
AK157079.1 | 4 | 477 | BAE33956.1 | |
BC119511.1 | 4 | 424 | ||
BC119512.1 | 4 | 424 | ||
X53798.1 | 4 | 450 | CAA37807.1 |
More Information
Set Membership: |
Intragenic Exonic |