Assay Details
Target Gene Details
Entrez Gene ID: | 109979 |
Gene Name: | ADP-ribosyltransferase 3 |
Gene Aliases: |
4930569O04Rik, ARTC3 |
Location: |
Chr.5:92331799-92414628 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Art3 | NM_181728.2 | NP_859417.2 | ||
AK016257.1 | ||||
DQ519368.1 | ||||
DQ519369.1 | ||||
DQ810295.1 |
Target Gene Details
Entrez Gene ID: | 15945 |
Gene Name: | chemokine (C-X-C motif) ligand 10 |
Gene Aliases: |
C7, CRG-2, INP10, IP-10, IP10, Ifi10, Scyb10, gIP-10, mob-1 |
Location: |
Chr.5:92346638-92348889 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cxcl10 | NM_021274.2 | 4 | 598 | NP_067249.1 |
AF227743.1 | 4 | 559 | AAG10400.1 | |
AK144279.1 | 4 | 589 | BAE25810.1 | |
AK146144.1 | 4 | 596 | BAE26931.1 | |
AK150380.1 | 4 | 589 | BAE29511.1 | |
AK150765.1 | 4 | 589 | BAE29832.1 | |
AK151210.1 | 4 | 589 | BAE30205.1 | |
AK151248.1 | 4 | 589 | BAE30238.1 | |
AK151415.1 | 4 | 589 | BAE30380.1 | |
AK151534.1 | 4 | 589 | BAE30481.1 | |
AK152568.1 | 4 | 589 | BAE31320.1 | |
AK152814.1 | 4 | 589 | BAE31517.1 | |
AK152838.1 | 1 | 97 | ||
AK152924.1 | 3 | 431 | BAE31599.1 | |
AK153181.1 | 4 | 589 | BAE31783.1 | |
AK157130.1 | 4 | 589 | BAE33973.1 | |
AK157139.1 | 4 | 589 | BAE33975.1 | |
AK157589.1 | 4 | 589 | BAE34130.1 | |
AK157678.1 | 4 | 589 | BAE34149.1 | |
AK172540.1 | 4 | 589 | BAE43058.1 | |
BC030067.1 | 4 | 565 | AAH30067.1 | |
M33266.1 | 4 | 563 | AAA02968.1 | |
M86829.1 | 4 | 588 | AAA21489.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |