Assay Details
Target Gene Details
Entrez Gene ID: | 67460 |
Gene Name: | 2,4-dienoyl CoA reductase 1, mitochondrial |
Gene Aliases: |
1200012F07Rik, Decr, Nadph |
Location: |
Chr.4:15917240-15945507 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Decr1 | NM_026172.3 | 10 | 1416 | NP_080448.1 |
AK004725.2 | 10 | 1283 | BAB23508.1 | |
AK146003.1 | 10 | 1287 | BAE26824.1 | |
BC046972.1 | 10 | 1416 | AAH46972.1 |
More Information
Set Membership: |
Intragenic Exonic |