Assay Details
Target Gene Details
Entrez Gene ID: | 73314 |
Gene Name: | leucine rich repeat containing 69 |
Gene Aliases: |
1700034K16Rik |
Location: |
Chr.4:14623622-14801320 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lrrc69 | NM_028499.2 | 8 | 1032 | NP_082775.1 |
AK006601.1 | BAB24669.1 | |||
AK161371.1 | 8 | 1035 | BAE36353.1 |
More Information
Set Membership: |
Intragenic Exonic |