Assay Details
Target Gene Details
Entrez Gene ID: | 104382 |
Gene Name: | BarH-like 2 (Drosophila) |
Gene Aliases: |
E130309B19Rik, MBH1 |
Location: |
Chr.5:106452523-106458166 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Barhl2 | NM_001005477.1 | 3 | 1555 | NP_001005477.1 |
AK053799.1 | 3 | 1555 | ||
BC055789.1 | 4 | 868 | AAH55789.1 | |
BC078444.1 | 3 | 1273 | AAH78444.1 |
More Information
Set Membership: |
Intragenic Exonic |