Assay Details
Target Gene Details
Entrez Gene ID: | 14073 |
Gene Name: | fatty acid amide hydrolase |
Gene Aliases: |
AW412498 |
Location: |
Chr.4:115996656-116017902 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Faah | NM_010173.4 | 15 | 2184 | NP_034303.3 |
AK004985.1 | 14 | 2708 | ||
AK079598.1 | 15 | 2185 | BAC37695.1 | |
AK080603.1 | 15 | 2017 | ||
AK138481.1 | 15 | 1804 | BAE23677.1 | |
BC006863.1 | 15 | 1963 | AAH06863.1 | |
BC052321.1 | 15 | 2226 | AAH52321.1 | |
U82536.1 | AAB58506.1 |
More Information
Set Membership: |
Intragenic Exonic |