Assay Details
Target Gene Details
Entrez Gene ID: | 21687 |
Gene Name: | endothelial-specific receptor tyrosine kinase |
Gene Aliases: |
AA517024, Cd202b, Hyk, STK1, Tie-2, Tie2 |
Location: |
Chr.4:94739289-94874976 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tek | NM_001290549.1 | 23 | 4314 | NP_001277478.1 |
NM_001290551.1 | 22 | 4164 | NP_001277480.1 | |
NM_013690.3 | 23 | 4317 | NP_038718.2 | |
BC050824.1 | 22 | 4158 | AAH50824.1 | |
D13738.1 | 23 | 4322 | BAA02883.1 | |
S67051.1 | 24 | 3977 | AAB28663.1 | |
X67553.1 | 23 | 4085 | CAA47857.1 |
More Information
Set Membership: |
Intragenic Exonic |