Assay Details
Target Gene Details
Entrez Gene ID: | 230025 |
Gene Name: | PR domain containing 13 |
Gene Aliases: |
- |
Location: |
Chr.4:21675865-21685963 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prdm13 | NM_001080771.1 | 1 | 157 | NP_001074240.1 |
AK149252.1 | ||||
BB665287.1 |
More Information
Set Membership: |
Intragenic Exonic |