Assay Details
Target Gene Details
Entrez Gene ID: | 22138 |
Gene Name: | titin |
Gene Aliases: |
1100001C23Rik, 2310036G12Rik, 2310057K23Rik, 2310074I15Rik, AF006999, AV006427, D330041I19Rik, D830007G01Rik, L56, mdm, shru |
Location: |
Chr.2:76703980-76982557 on Build GRCm38 |
Assay Gene Location: | Within Exon 290 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ttn | NM_011652.3 | 289 | 87074 | NP_035782.3 |
NM_028004.2 | 168 | 67331 | NP_082280.2 |