Assay Details
Target Gene Details
Entrez Gene ID: | 53975 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 20 |
Gene Aliases: |
GEMIN3, dp103 |
Location: |
Chr.3:105678270-105688113 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx20 | NM_017397.3 | 11 | 1676 | NP_059093.3 |
AF217000.1 | 11 | 1551 | AAK56847.1 | |
AF217001.1 | AAK56848.1 | |||
AF220454.1 | 11 | 1679 | AAF76301.1 | |
AJ250027.1 | 11 | 1551 | CAB86201.1 | |
AK154518.1 | 11 | 1555 | ||
BC125471.1 | 11 | 1676 | ||
BC137721.1 | 11 | 1676 |