Assay Details
Target Gene Details
Entrez Gene ID: | 19338 |
Gene Name: | RAB33B, member RAS oncogene family |
Gene Aliases: |
- |
Location: |
Chr.3:51483966-51496228 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rab33b | NM_016858.2 | 2 | 1575 | NP_058554.1 |
AB004665.1 | 2 | 1525 | BAA22655.1 | |
AK004974.1 | 2 | 1311 | BAB23710.1 | |
AK087967.1 | 2 | 1315 | BAC40064.1 | |
AK139821.1 | 2 | 1561 | BAE24147.1 | |
AK144706.1 | 2 | 1313 | ||
AK159674.1 | 2 | 1310 | BAE35278.1 | |
AK163783.1 | 2 | 1562 | BAE37491.1 | |
AK170483.1 | 2 | 1204 | BAE41825.1 | |
BC065076.1 | 2 | 1575 | AAH65076.1 |