Assay Details
Target Gene Details
Entrez Gene ID: | 213262 |
Gene Name: | follistatin-like 5 |
Gene Aliases: |
9130207J01Rik |
Location: |
Chr.3:76074275-76710010 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fstl5 | NM_001253719.1 | 16 | 2427 | NP_001240648.1 |
NM_178673.4 | 16 | 2687 | NP_848788.2 | |
AF374460.1 | 16 | 2661 | AAQ02778.1 | |
AK033652.1 | 16 | 2678 | BAC28408.1 | |
AK046070.1 | 16 | 2428 | BAC32590.1 | |
AK081152.1 | 16 | 2678 | BAC38146.1 | |
AK122482.1 | 7 | 1056 | BAC65764.1 | |
AK138388.1 | 16 | 2979 | BAE23641.1 | |
BC075640.1 | 16 | 2650 | AAH75640.1 |
More Information
Set Membership: |
Intragenic Exonic |