Assay Details
Target Gene Details
Entrez Gene ID: | 114249 |
Gene Name: | nephronectin |
Gene Aliases: |
1110009H02Rik, AA682063, AI314031, Nctn, POEM |
Location: |
Chr.3:132881745-132950291 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Npnt | NM_001029836.2 | 12 | 2155 | NP_001025007.1 |
NM_001287101.1 | 14 | 2299 | NP_001274030.1 | |
NM_001287102.1 | 13 | 2248 | NP_001274031.1 | |
NM_001287103.1 | 12 | 1873 | NP_001274032.1 | |
NM_033525.3 | 13 | 2206 | NP_277060.2 | |
AB059656.1 | 12 | 2155 | BAB69692.1 | |
AK050484.1 | 12 | 2059 | BAC34283.1 | |
AK158764.1 | 12 | 1875 | BAE34649.1 | |
AY035898.1 | 12 | 2030 | AAK96010.1 | |
AY035899.1 | 13 | 2081 | AAK96011.1 | |
BC046642.1 | 7 | 1342 | AAH46642.1 | |
BC068308.1 | 12 | 2024 | AAH68308.1 |