Assay Details
Target Gene Details
Entrez Gene ID: | 20732 |
Gene Name: | serine protease inhibitor, Kunitz type 1 |
Gene Aliases: |
HAI-1 |
Location: |
Chr.2:119237360-119249535 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Spint1 | NM_016907.3 | 2 | 293 | NP_058603.2 |
AF099018.1 | 2 | 248 | AAF02490.1 | |
AK017342.1 | 1 | 175 | BAB30697.1 | |
BC005769.1 | 1 | 180 | AAH05769.1 |
More Information
Set Membership: |
Intragenic Exonic |