Assay Details
Target Gene Details
Entrez Gene ID: | 11876 |
Gene Name: | artemin |
Gene Aliases: |
neublastin |
Location: |
Chr.4:117926160-117930791 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Artn | NM_001284191.1 | 1 | 64 | NP_001271120.1 |
NM_001284192.1 | 1 | 64 | NP_001271121.1 | |
NM_001284193.1 | 1 | 64 | NP_001271122.1 | |
NM_009711.4 | 1 | 64 | NP_033841.1 | |
AK053914.1 | 1 | 65 | BAC35590.1 |
More Information
Set Membership: |
Intragenic Exonic |