Assay Details
Target Gene Details
Entrez Gene ID: | 192167 |
Gene Name: | neuroligin 1 |
Gene Aliases: |
6330415N05Rik, BB179718, NL1, Nlg1, mKIAA1070 |
Location: |
Chr.3:25431841-26332056 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nlgn1 | NM_001163387.1 | 5 | 1848 | NP_001156859.1 |
NM_138666.3 | 7 | 2875 | NP_619607.2 | |
AK122433.1 | 7 | 2876 | BAC65715.1 | |
AK138209.1 | 2 | 709 | BAE23583.1 | |
BC098461.1 | 5 | 1848 | AAH98461.1 |
More Information
Set Membership: |
Intragenic Exonic |