Assay Details
Target Gene Details
Entrez Gene ID: | 12972 |
Gene Name: | crystallin, zeta |
Gene Aliases: |
Sez9 |
Location: |
Chr.3:154596711-154623182 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cryz | NM_009968.3 | 1 | 526 | NP_034098.1 |
AK098082.1 | ||||
AK133820.1 | 1 | 527 | ||
AK134281.1 | BAE22081.1 | |||
BB618817.1 | 1 | 527 | ||
BC003800.1 | AAH03800.1 | |||
BC043076.1 | AAH43076.1 | |||
S70056.1 | AAB30620.2 |
More Information
Set Membership: |
Intragenic Exonic |