Assay Details
Target Gene Details
Entrez Gene ID: | 20672 |
Gene Name: | SRY (sex determining region Y)-box 18 |
Gene Aliases: |
AI385749, Ra, Ragl |
Location: |
Chr.2:181669837-181671640 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sox18 | NM_009236.2 | 1 | 237 | NP_033262.2 |
AK144281.1 | 1 | 238 | BAE25811.1 | |
BC006612.1 | 1 | 237 | AAH06612.1 | |
BY141110.1 | 1 | 233 | ||
L35032.1 | 1 | 206 | AAB18615.1 |
More Information
Set Membership: |
Intragenic Exonic |