Assay Details
Target Gene Details
Entrez Gene ID: | 22138 |
Gene Name: | titin |
Gene Aliases: |
1100001C23Rik, 2310036G12Rik, 2310057K23Rik, 2310074I15Rik, AF006999, AV006427, D330041I19Rik, D830007G01Rik, L56, mdm, shru |
Location: |
Chr.2:76703980-76982557 on Build GRCm38 |
Assay Gene Location: | Within Exon 314 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ttn | NM_011652.3 | 313 | 101236 | NP_035782.3 |
NM_028004.2 | 192 | 81493 | NP_082280.2 | |
AK003152.1 | 4 | 1651 | BAB22606.1 | |
AK020463.1 | 1 | 570 | ||
AK029024.1 | 6 | 2334 | BAC26249.1 | |
BC025840.1 | 3 | 1684 | AAH25840.1 |