Assay Details
Target Gene Details
Entrez Gene ID: | 230163 |
Gene Name: | aldolase B, fructose-bisphosphate |
Gene Aliases: |
Aldo-2, Aldo2, BC016435 |
Location: |
Chr.4:49535995-49549483 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aldob | NM_144903.2 | 9 | 1650 | NP_659152.1 |
AK149395.1 | 9 | 1667 | BAE28850.1 | |
AK167566.1 | 9 | 1664 | BAE39629.1 | |
BC024056.1 | 9 | 1656 | AAH24056.1 | |
BC036132.1 | 9 | 1650 | AAH36132.1 |
More Information
Set Membership: |
Intragenic Exonic |