Assay Details
Target Gene Details
Entrez Gene ID: | 68259 |
Gene Name: | intraflagellar transport 80 |
Gene Aliases: |
4921524P20Rik, Wdr56, mKIAA1374 |
Location: |
Chr.3:68892499-69004653 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ift80 | NM_026641.2 | 20 | 3816 | NP_080917.1 |
AK077870.1 | 1 | 1384 | BAC37042.1 | |
AK147305.1 | 19 | 7343 | ||
BC013814.1 | 15 | 3105 | AAH13814.1 | |
BC034101.1 | 20 | 3711 | AAH34101.1 |