Assay Details
Target Gene Details
Entrez Gene ID: | 20713 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade I, member 1 |
Gene Aliases: |
AI837402, Ns, PI-12, PI12, Spi17 |
Location: |
Chr.3:75557503-75642523 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Serpini1 | NM_009250.2 | 9 | 2203 | NP_033276.1 |
AJ001700.1 | 9 | 2172 | CAA04939.1 | |
AK032152.1 | 9 | 2192 | BAC27727.1 | |
AK133770.1 | 9 | 2193 |
More Information
Set Membership: |
Intragenic Exonic |